Ohaus® Power Adapters
Replacement power adapter and plugs
Ohaus Prower Adapter, model 11003-55 is a 120 VAC, 50/60 Hz adapter. This adapter does not include plug. Must order plug kit 11611-01 separately that includes 4 plugs (EU-US-AU-UK). Compatible with Ohaus series Scout Pro SP, SD, Catapult C11, CL, CS, ES and Navigator NVx balances.
6,800.00 - ₹15,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Ohaus Power Adapter Prower Adapter; 120 V, 50/60 Hz WW-11003-55 Mfr # 46001724
COMPARE
115
₹15,800.00INR / Each
Ohaus 46001780 Plug Set includes Plug Adaptors for Au/Eu/Gb/Us WW-11611-01 Mfr # 46001780
COMPARE
₹6,800.00INR / Each