Ohaus Power Adapter Prower Adapter; 120 V, 50/60 Hz

Representative image only.

Ohaus Power Adapter Prower Adapter; 120 V, 50/60 Hz

Ohaus | Mfr: 46001724| Item: 1100355

Replacement power adapter and plugs

₹20,800.00INR / Each

₹20,800.00INR / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Ohaus Prower Adapter, model 11003-55 is a 120 VAC, 50/60 Hz adapter. This adapter does not include plug. Must order plug kit 11611-01 separately that includes 4 plugs (EU-US-AU-UK). Compatible with Ohaus series Scout Pro SP, SD, Catapult C11, CL, CS, ES and Navigator NVx balances.

Ohaus Prower Adapter; 120 VAC, 50/60 Hz. Adapter does not include plug. Must order plug kit p/n 11611-01 that includes 4 plugs (EU-US-AU-UK)

1 Year Warranty
Specifications
  • Power (VAC)115
  • DescriptionPrower Adapter; 120 V, 50/60 Hz
  • Warranty1 year
Reviews
Q&A