Ohaus 46001780 Plug Set includes Plug Adaptors for Au/Eu/Gb/Us
Representative image only.
Ohaus 46001780 Plug Set includes Plug Adaptors for Au/Eu/Gb/Us
Ohaus | Mfr: 46001780| Item: 1161101
Replacement power adapter and plugs
₹9,300.00INR / Each
₹9,300.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Ohaus Prower Adapter, model 11003-55 is a 120 VAC, 50/60 Hz adapter. This adapter does not include plug. Must order plug kit 11611-01 separately that includes 4 plugs (EU-US-AU-UK). Compatible with Ohaus series Scout Pro SP, SD, Catapult C11, CL, CS, ES and Navigator NVx balances.
1 Year Warranty
Specifications
  • LanguagesEnglish
  • Height (in)2
  • Width (in)4
  • Depth (in)5
  • DescriptionPlug Setincludes Plug Adaptors for Au/Eu/Gb/Us