Ohaus® Pioneer® PX Analytical Balances
Outstanding accuracy and repeatability at an affordable price
  • Bright backlit dual-line LCD is easy to view in any environment
  • Durable construction with metal base, ABS top housing, and stainless steel weighing pan
  • Standard RS-232 and USB ports for easy connectivity to printers and other devices
  • Four quick keys allow for easy operation of multiple application modes
  • Static removal bar provides convenient grounding
The Pioneer® PX analytical balances deliver high weighing accuracy and repeatability at an affordable price for use in laboratory, industrial, and education applications. A bright backlit LCD provides a clear view of weighing results while a second line display offers additional information and guidance.
A static removal bar located at the top of the draft shield discharges any accumulated static charge to increase accuracy in weighing results. The draft shield features glass panels that can be removed and replaced for fast, simple cleaning.
Standard RS-232 and USB ports make transferring results to a PC or other peripherals a simple process. The balances are equipped with a security lock to prevent theft. A standard in-use cover provides added protection against spillage and minimizes damage to the display and keypad. Other features include four user selectable operating languages (English, Chinese, Japanese, and Korean), real time clock, GMP/GLP data fields, level indicator, and menu lock. Application modes include basic weighing, parts counting, percent weighing, animal weighing, and density determination.
309,000.00 - ₹909,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Ohaus PX84/E Pioneer Analytical Balance, 84g x 0.0001g, External Calibration with Draft Shield 1161175 Mfr # PX84/E
COMPARE
External 84 0.0001
₹531,000.00INR / Each
Ohaus PX84 Pioneer Analytical Balance, 84g x 0.0001g, Internal Calibration with Draft Shield 1161176 Mfr # PX84
COMPARE
Internal 84 0.0001
₹620,000.00INR / Each
Ohaus PX124/E Pioneer Analytical Balance, 120g x 0.0001g, External Calibration with Draft Shield 1161177 Mfr # PX124/E
COMPARE
External 120 0.0001
₹613,000.00INR / Each
Ohaus PX124  Pioneer Analytical Balance, 120g x 0.0001g, Internal Calibration with Draft Shield 1161178 Mfr # PX124
COMPARE
Internal 120 0.0001
₹709,000.00INR / Each
Ohaus PX224/E Pioneer Analytical Balance, 220g x 0.0001g, External Calibration with Draft Shield 1161179 Mfr # PX224/E
COMPARE
External 220 0.0001
₹680,000.00INR / Each
Ohaus PX224 Pioneer Analytical Balance, 220g x 0.0001g, Internal Calibration with Draft Shield 1161180 Mfr # PX224
COMPARE
Internal 220 0.0001
₹909,000.00INR / Each
Ohaus PX163/E Pioneer Analytical Balance, 160g x 0.001g, External Calibration with Draft Shield 1161181 Mfr # PX163/E
COMPARE
External 160 0.001
₹309,000.00INR / Each
Ohaus PX163 Pioneer Analytical Balance, 160g x 0.001g, Internal Calibration with Draft Shield 1161182 Mfr # PX163
COMPARE
Internal 160 0.001
₹453,000.00INR / Each
Ohaus PX323/E Pioneer Analytical Balance, 320g x 0.001g, External Calibration with Draft Shield 1161183 Mfr # PX323/E
COMPARE
External 320 0.001
₹420,000.00INR / Each
Ohaus PX323 Pioneer Analytical Balance, 320g x 0.001g, Internal Calibration with Draft Shield 1161184 Mfr # PX323
COMPARE
Internal 320 0.001
₹436,000.00INR / Each
Showing 1- 10 of 10