Ohaus® Explorer Semi-Micro Analytical Balances
The Explorer is perfect for university research, teaching laboratories, general lab and industrial applications
  • Modular design allows for the display to be separated from the weighing base for maximum flexibility in any work area
  • Optimized linearity and repeatability
  • AutoCal™ automatic internal calibration ensures results are accurate
  • USB host port lets you directly transfer weighing data to a flash drive
  • Intuitive operation with large color touch-screen display and icon-driven application software
  • Easy-to-use SmarText™ 2.0 graphical software
  • Touchless sensors for hands-free operation
The Ohaus Explorer’s advanced applications simplify even the most complex laboratory measurements. Whether it’s determining the difference between initial and residual weights or calculating the density of solids and liquids, these balances eliminate the need for time-consuming manual calculations and data logging. The high-resolution display and innovative user interface make balance setup and operation effortless.
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Ohaus EX125D Explorer Semi-Micro Dual Range Analytical Balance 120Gx0.1mg; 52gx0.01mg 1101548 Mfr # EX125D
COMPARE
120 0.1 Explorer Semi-Micro Dual Range Analytical Balance 120 g x 0.1 mg and 52 g x 0.01 mg
Call For Price
Ohaus EX125 Explorer Semi-Micro Analytical Balance 120Gx0.01mg 1101549 Mfr # EX125
COMPARE
120 0.01 Explorer Semi-Micro Analytical Balance 120 g x 0.01 mg
Call For Price
Ohaus EX225D Explorer Semi-Micro Dual Range Analytical Balance 220Gx0.1mg; 120Gx0.01mg 1101550 Mfr # EX225D
COMPARE
220 0.1 Explorer Semi-Micro Dual Range Analytical Balance 220 g x 0.1 mg and 120 g x 0.01 mg
Call For Price
Ohaus EX225/AD Explorer Semi-Micro Analytical Balance 220Gx0.01mg Automatic Door s 1101552 Mfr # EX225/AD
COMPARE
220 0.01 Explorer Semi-Micro Analytical Balance, 220 g x 0.01 mg; Automatic Door