Monarch Instrument Track-It™ RFID Temperature and Humidity Data Loggers with Android App
Wireless configuration and data transfer directly to your Android™ device preserves packaging
  • View data on your Android device or e-mail the data set for immediate remote review
  • Free app configures the record, programs sampling rate, and shows tabular and graphic views of data
  • Optional high-temperature battery (order separately) increases the upper limit of the logger from 60°C to 80°C
  • Optional docking station allows download to PC using the free Track-It™ software
Ideally suited for supply chain monitoring and similar applications, this data logger includes an RFID (radio frequency identification) antenna for communication with any NFC (near field communication) enabled Android device. The free app configures and starts/stops the data logging session, scans dataset for alarms, and shows the complete record in graphic, tabular, or summary form.
Affix the logger to the inside of a package or shipping carton and access the data through the package wall, protecting the integrity of the contents. Multiple user-programmable LED indicators signal normal logging and alarm states.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Monarch Track-It RFID RFID Temperature Data Logger 1800902 Mfr # 5396-2010
COMPARE
RFID Temperature Data Logger
Call For Price
Monarch Track-It RFID RFID Temperature and Humidity Data Logger 1800903 Mfr # 5396-2012
COMPARE
RFID Temperature and Humidity Data Logger