Mini-BeadBeater Glass Beads

Choose the bead material and size to suit your disruption application needs.
10,600.00 - ₹28,400.00INR / Pound of 1

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
BioSpec Products 11079110 Mini-BeadBeater Glass Mill Beads, 1.0 mm; 1 lb 3627050 Mfr # 11079110
COMPARE
Glass Mini-BeadBeater Glass Mill Beads, 1.0 mm; 1 lb
₹17,900.00INR / Pound of 1
BioSpec Products 11079105 Mini-BeadBeater Glass Mill Beads, 0.5 mm; 1 lb 3627051 Mfr # 11079105
COMPARE
Glass Mini-BeadBeater Glass Mill Beads, 0.5 mm; 1 lb
₹18,900.00INR / Pound of 1
BioSpec Products 11079101 Mini-BeadBeater Glass Mill Beads, 0.1 to 0.15 mm 3627052 Mfr # 11079101
COMPARE
Glass Mini-BeadBeater Glass Mill Beads, 0.1 to 0.15 mm
₹10,600.00INR / Pound of 1
BioSpec Products 11079105Z Mini-BeadBeater Zirconia-Silicate Beads, 0.5 mm; 1 lb 3627061 Mfr # 11079105Z
COMPARE
Zirconia-Silicate Mini-BeadBeater Zirconia-Silicate Beads, 0.5 mm; 1 lb
₹28,400.00INR / Pound of 1
BioSpec Products 11079101Z Mini-BeadBeater Zirconia-Silicate Beads, 0.1-0.15 mm; 1 lb 3627062 Mfr # 11079101Z
COMPARE
Zirconia-Silicate Mini-BeadBeater Zirconia-Silicate Beads, 0.1-0.15 mm; 1 lb
₹25,200.00INR / Pound of 1
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat