BioSpec Products 11079101Z Mini-BeadBeater Zirconia-Silicate Beads, 0.1-0.15 mm; 1 lb

Representative image only.

BioSpec Products 11079101Z Mini-BeadBeater Zirconia-Silicate Beads, 0.1-0.15 mm; 1 lb

BioSpec Products | Mfr: 11079101Z| Item: 3627062

₹25,200.00INR / Pound of 1

₹25,200.00INR / Pound of 1

Buy It Together

Current Item

BioSpec Products 11079101Z Mini-BeadBeater Zirconia-Silicate Beads, 0.1-0.15 mm; 1 lb
BioSpec Products 11079101Z Mini-BeadBeater Zirconia-Silicate Beads, 0.1-0.15 mm; 1 lb
3627062

₹25,200.00INR / Pound of 1
+
Cole-Parmer Glass Beads Refill, 3 mm, 1000 g
0185033

₹9,100.00INR / Each
=
Total Price 34300.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Choose the bead material and size to suit your disruption application needs.

1 Year Warranty
Specifications
  • MaterialZirconia-Silicate
  • DescriptionMini-BeadBeater Zirconia-Silicate Beads, 0.1-0.15 mm; 1 lb
  • Warranty1 Year
Reviews
Q&A