BioSpec Products 11079101 Mini-BeadBeater Glass Mill Beads, 0.1 to 0.15 mm

Representative image only.

BioSpec Products 11079101 Mini-BeadBeater Glass Mill Beads, 0.1 to 0.15 mm

BioSpec Products | Mfr: 11079101| Item: 3627052

₹10,600.00INR / Pound of 1

₹10,600.00INR / Pound of 1

Buy It Together

Current Item

BioSpec Products 11079101 Mini-BeadBeater Glass Mill Beads, 0.1 to 0.15 mm
BioSpec Products 11079101 Mini-BeadBeater Glass Mill Beads, 0.1 to 0.15 mm
3627052

₹10,600.00INR / Pound of 1
+
Cole-Parmer Glass Beads Refill, 3 mm, 1000 g
0185033

₹9,100.00INR / Each
=
Total Price 19700.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Choose the bead material and size to suit your disruption application needs.

1 Year Warranty
Specifications
  • MaterialGlass
  • DescriptionMini-BeadBeater Glass Mill Beads, 0.1 to 0.15 mm
  • Warranty1 Year
Reviews
Q&A