Microbial and Iron Test Kits
Make sure surfaces are contamination-free
  • Strips are flexible-sample in hard-to-reach areas
  • Quick check for bacteria, yeat, and molds to prevent corrosion, slime, formation, and odors
  • Kits are complete and ready to use - no special equipment or training necessary
Kits contain flexible plastic strips to which filter paper containing nutrients are attached. Aerobic bacteria form red spots on the test strip. Yeast and mold form black spots on the test strip. Use the number of spots along with the degree of background reddening for the aerobic bacteria to get a measurement of microbial growth.
For the yeast and mold, use the number of spots along with the degree of graying of the background to get a measurement of yeast and mold growth. After an incubation period, compare the strip to a color chart on the label for an estimated bacterial count.
4,400.00 - ₹37,500.00INR / Pkg of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test
Availability Pricing
COMPARE
Bacteria, aerobic
₹37,500.00INR / Pkg of 1
COMPARE
Fungi, yeasts and molds
₹37,200.00INR / Pkg of 25
ITS 480025 Iron test strips (0, 0.02, 0.05, 0.1, 0.2, 0.3, 0.5, 0.75, 1.0, 2.0, 5.0 ppm) 9953126 Mfr # 480025
COMPARE
0, 0.02, 0.05, 0.1, 0.2, 0.3, 0.5, 0.75, 1.0, 2.0, 5.0 ppm Iron
₹4,400.00INR / Each