Cole-Parmer Microbial Test Kits, Aerobic Bacteria Test Kit
Representative image only.
Cole-Parmer Microbial Test Kits, Aerobic Bacteria Test Kit
Cole-Parmer | Item: 1402020
Make sure surfaces are contamination-free
₹37,500.00INR / Pkg of 1
₹37,500.00INR / Pkg of 1
Buy It Together
Current Item
Cole-Parmer Microbial Test Kits, Aerobic Bacteria Test Kit
Cole-Parmer Microbial Test Kits, Aerobic Bacteria Test Kit
1402020
₹37,500.00INR / Pkg of 1
+
Environmental Express Starter Kit, Basic Mold Sampling; 120V
6006203
₹102,000.00INR / Each
=
Total Price 139500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Strips are flexible-sample in hard-to-reach areas
  • Quick check for bacteria, yeat, and molds to prevent corrosion, slime, formation, and odors
  • Kits are complete and ready to use - no special equipment or training necessary
More About this Item
Kits contain flexible plastic strips to which filter paper containing nutrients are attached. Aerobic bacteria form red spots on the test strip. Yeast and mold form black spots on the test strip. Use the number of spots along with the degree of background reddening for the aerobic bacteria to get a measurement of microbial growth.
For the yeast and mold, use the number of spots along with the degree of graying of the background to get a measurement of yeast and mold growth. After an incubation period, compare the strip to a color chart on the label for an estimated bacterial count.
Specifications
  • TestBacteria, aerobic
  • Reagent PlatformStrips
  • Tests Per Kit25
  • DescriptionMicrobial Test Kits, Aerobic Bacteria Test Kit