ITS 480025 Iron test strips (0, 0.02, 0.05, 0.1, 0.2, 0.3, 0.5, 0.75, 1.0, 2.0, 5.0 ppm)

Representative image only.

ITS 480025 Iron test strips (0, 0.02, 0.05, 0.1, 0.2, 0.3, 0.5, 0.75, 1.0, 2.0, 5.0 ppm)

Cole-Parmer | Mfr: 480025| Item: 9953126

Make sure surfaces are contamination-free

₹4,400.00INR / Each

₹4,400.00INR / Each

Buy It Together

Current Item

ITS 480025 Iron test strips (0, 0.02, 0.05, 0.1, 0.2, 0.3, 0.5, 0.75, 1.0, 2.0, 5.0 ppm)
ITS 480025 Iron test strips (0, 0.02, 0.05, 0.1, 0.2, 0.3, 0.5, 0.75, 1.0, 2.0, 5.0 ppm)
9953126

₹4,400.00INR / Each
+
Environmental Express Starter Kit, Basic Mold Sampling; 120V
6006203

₹102,000.00INR / Each
=
Total Price 106400.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Strips are flexible-sample in hard-to-reach areas
  • Quick check for bacteria, yeat, and molds to prevent corrosion, slime, formation, and odors
  • Kits are complete and ready to use - no special equipment or training necessary

More About this Item

Kits contain flexible plastic strips to which filter paper containing nutrients are attached. Aerobic bacteria form red spots on the test strip. Yeast and mold form black spots on the test strip. Use the number of spots along with the degree of background reddening for the aerobic bacteria to get a measurement of microbial growth.

For the yeast and mold, use the number of spots along with the degree of graying of the background to get a measurement of yeast and mold growth. After an incubation period, compare the strip to a color chart on the label for an estimated bacterial count.

Specifications
  • TestIron
  • Measurement Points0, 0.02, 0.05, 0.1, 0.2, 0.3, 0.5, 0.75, 1.0, 2.0, 5.0 ppm
  • Reagent PlatformStrips
  • Tests Per Kit25
  • DescriptionIron test strips (0, 0.02, 0.05, 0.1, 0.2, 0.3, 0.5, 0.75, 1.0, 2.0, 5.0 ppm)
Reviews
Q&A