Mettler Toledo® Seven2Go™ Conductivity Meters
Reliable, simple design for one-handed, portable use
  • Take readings quickly with one hand
  • Don't recalibrate with every electrode change—compatible with ISM sensors
  • Robust IP67-rated housing protects meter in harsh environments
These handheld conductivity meters are perfect for mobile applications. Rugged construction provides durability, while the navigation pad presents an intuitive interface and T-pad button configuration.
The InLab ISM (Intelligent Sensor Management) series of electrodes stores its ID number, serial number, and calibration data. Connect the conductivity cell to the meter it and it automatically communicates this information—no need to select electrode type when switching or recalibrate every time you power on the meter.
Choose the meter alone if you already have ISM electrodes, or choose a bundled kit suited to your application.
186,000.00 - ₹434,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo S3-Meter Portable Conductivity Meter; 0.01 uS to 500 mS 3590350 Mfr # 30207954
COMPARE
Portable Conductivity Meter; 0.01 uS to 500 mS
₹186,000.00INR / Each
Mettler Toledo S3-Standard Kit Portable Conductivity Meter; Standard Kit 3590351 Mfr # 30207955
COMPARE
Portable Conductivity Meter; Standard Kit
₹270,000.00INR / Each
Mettler Toledo S3-Field Kit Portable Conductivity Meter; Field Kit 3590352 Mfr # 30207956
COMPARE
Portable Conductivity Meter; Field Kit
₹287,000.00INR / Each
Mettler Toledo S7-Meter Portable Conductivity Meter; Pro, 0.01 uS to 1000 mS 3590357 Mfr # 30207961
COMPARE
Portable Conductivity Meter; Pro, 0.01 uS to 1000 mS
₹298,000.00INR / Each
Mettler Toledo S7-Standard Kit Portable Conductivity Meter; Pro Standard Kit 3590358 Mfr # 30207962
COMPARE
Portable Conductivity Meter; Pro Standard Kit
₹394,000.00INR / Each
Mettler Toledo S7-Field Kit Portable Conductivity Meter; Pro Field Kit 3590359 Mfr # 30207963
COMPARE
Portable Conductivity Meter; Pro Field Kit
₹410,000.00INR / Each
Mettler Toledo S7-USP/EP Kit Portable Conductivity Meter Kit; Pro USP/EP Kit 3590360 Mfr # 30207873
COMPARE
Portable Conductivity Meter Kit; Pro USP/EP Kit
₹434,000.00INR / Each
Showing 1- 7 of 7