Mettler Toledo S3-Standard Kit Portable Conductivity Meter; Standard Kit
Representative image only.
Mettler Toledo S3-Standard Kit Portable Conductivity Meter; Standard Kit
Mettler Toledo | Mfr: 30207955| Item: 3590351
Reliable, simple design for one-handed, portable use
₹270,000.00INR / Each
₹270,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo S3-Standard Kit Portable Conductivity Meter; Standard Kit
Mettler Toledo S3-Standard Kit Portable Conductivity Meter; Standard Kit
3590351
₹270,000.00INR / Each
+ =
Total Price 541000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Meter, InLab® 738-ISM probe (35902-46), electrode clip, calibration solution pouches (1413 µS and 12.88 mS), and batteries.
Key Features
  • Take readings quickly with one hand
  • Don't recalibrate with every electrode change—compatible with ISM sensors
  • Robust IP67-rated housing protects meter in harsh environments
  • 1 year warranty
More About this Item
These handheld conductivity meters are perfect for mobile applications. Rugged construction provides durability, while the navigation pad presents an intuitive interface and T-pad button configuration.
The InLab ISM (Intelligent Sensor Management) series of electrodes stores its ID number, serial number, and calibration data. Connect the conductivity cell to the meter it and it automatically communicates this information—no need to select electrode type when switching or recalibrate every time you power on the meter.
Choose the meter alone if you already have ISM electrodes, or choose a bundled kit suited to your application.
1 Year WarrantyIP 67
Specifications
  • DescriptionPortable Conductivity Meter; Standard Kit