Mettler Toledo Laboratory Conductivity Cells
Outstanding performance for any application
  • Integrated temperature compensation sensor ensures measurements are accurate
  • Choose from a variety of materials and cable lengths to suit your requirements
These laboratory conductivity cells are all dip style with shaft materials ranging from plastic to metal to glass, and graphite, stainless steel, or platinum electrode bands. Common shaft diameter of 12 mm is compatible with most electrode holders. Rugged IP67-rated cells are preferred for outdoor and industrial use.
Intelligent Sensor Management (ISM) conductivity cells feature onboard electronic memory that stores calibration data, unique identification number, and temperature history. When connected to your compatible conductivity meter, the sensor is automatically recognized and the stored data uploads to the meter for immediate use, saving time and preventing errors.
102,000.00 - ₹325,000.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo 731 Conductivity Cell, standard range; 1.2 m cable WW-35902-40 Mfr # 51344020
COMPARE
Conductivity Cell, standard range; 1.2 m cable
₹118,000.00INR / Each
Mettler Toledo 731-ISM Conductivity Cell, standard range, ISM; 1.2 m cable WW-35902-41 Mfr # 30014092
COMPARE
Conductivity Cell, standard range, ISM; 1.2 m cable
₹129,000.00INR / Each
Mettler Toledo 731-2M Conductivity Cell, standard range; 2 m cable WW-35902-42 Mfr # 51344022
COMPARE
Conductivity Cell, standard range; 2 m cable
₹126,000.00INR / Each
Mettler Toledo 731-ISM-2M Conductivity Cell, standard range, ISM; 2 m cable WW-35902-43 Mfr # 30014093
COMPARE
Conductivity Cell, standard range, ISM; 2 m cable
₹135,000.00INR / Each
Mettler Toledo 738 Conductivity Cell, standard range, IP67; 1.8 m cable WW-35902-44 Mfr # 51344120
COMPARE
Conductivity Cell, standard range, IP67; 1.8 m cable
₹121,000.00INR / Each
Mettler Toledo 738-ISM Conductivity Cell, standard range, IP67, ISM; 1.8 m cable WW-35902-46 Mfr # 51344110
COMPARE
Conductivity Cell, standard range, IP67, ISM; 1.8 m cable
₹147,000.00INR / Each
Mettler Toledo 738-ISM-5M Conductivity Cell, standard range, IP67, ISM; 5 m cable WW-35902-47 Mfr # 51344112
COMPARE
Conductivity Cell, standard range, IP67, ISM; 5 m cable
₹176,000.00INR / Each
Mettler Toledo 741 Conductivity Cell, low range; 1.2 m cable WW-35902-49 Mfr # 51344024
COMPARE
Conductivity Cell, low range; 1.2 m cable
₹177,000.00INR / Each
Mettler Toledo 741-ISM Conductivity Cell, low range, ISM; 1.2 m cable WW-35902-50 Mfr # 30014094
COMPARE
Conductivity Cell, low range, ISM; 1.2 m cable
₹184,000.00INR / Each
Mettler Toledo 742 Conductivity Cell, low range, IP67; 1.8 m cable WW-35902-53 Mfr # 51344126
COMPARE
Conductivity Cell, low range, IP67; 1.8 m cable
₹176,000.00INR / Each
Mettler Toledo 742-5M Conductivity Cell, low range, IP67; 5 m cable WW-35902-54 Mfr # 51344128
COMPARE
Conductivity Cell, low range, IP67; 5 m cable
₹186,000.00INR / Each
Mettler Toledo 742-ISM Conductivity Cell, low range, IP67, ISM; 1.8 m cable WW-35902-55 Mfr # 51344116
COMPARE
Conductivity Cell, low range, IP67, ISM; 1.8 m cable
₹188,000.00INR / Each
Mettler Toledo Trace Digital Conductivity Cell, low range; 1.8 m cable WW-35902-57 Mfr # 30014097
COMPARE
Digital Conductivity Cell, low range; 1.8 m cable
₹325,000.00INR / Each
Mettler Toledo 710 Conductivity Cell, standard range, aggressive / organics; 1.2 m cable WW-35902-60 Mfr # 51302256
COMPARE
Conductivity Cell, standard range, aggressive / organics; 1.2 m cable
₹102,000.00INR / Each
Mettler Toledo 751-4MM Conductivity Cell, small sample, 4 mm dia; 1.2 m cable WW-35902-63 Mfr # 51344030
COMPARE
Conductivity Cell, small sample, 4 mm dia; 1.2 m cable
₹194,000.00INR / Each
Mettler Toledo 752-6MM Conductivity Cell, small sample, 6 mm dia; 1.2 m cable WW-35902-64 Mfr # 51344031
COMPARE
Conductivity Cell, small sample, 6 mm dia; 1.2 m cable
₹177,000.00INR / Each
Showing 1- 16 of 16