Mettler Toledo S7-Field Kit Portable Conductivity Meter; Pro Field Kit
Representative image only.
Mettler Toledo S7-Field Kit Portable Conductivity Meter; Pro Field Kit
Mettler Toledo | Mfr: 30207963| Item: 3590359
Reliable, simple design for one-handed, portable use
₹410,000.00INR / Each
₹410,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo S7-Field Kit Portable Conductivity Meter; Pro Field Kit
Mettler Toledo S7-Field Kit Portable Conductivity Meter; Pro Field Kit
3590359
₹410,000.00INR / Each
+ =
Total Price 681000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Meter, LabX direct pH, InLab® 738-ISM probe (35902-46), electrode clip, calibration solution bottles (1413 µS and 12.88 mS), carrying case, USB cable, and batteries.
Key Features
  • Take readings quickly with one hand
  • Don't recalibrate with every electrode change—compatible with ISM sensors
  • Robust IP67-rated housing protects meter in harsh environments
  • Pro meters and kits add 12 languages, sunlight-proof display, and LED status lights
  • 1 year warranty
More About this Item
These handheld conductivity meters are perfect for mobile applications. Rugged construction provides durability, while the navigation pad presents an intuitive interface and T-pad button configuration.
The InLab ISM (Intelligent Sensor Management) series of electrodes stores its ID number, serial number, and calibration data. Connect the conductivity cell to the meter it and it automatically communicates this information—no need to select electrode type when switching or recalibrate every time you power on the meter.
Choose the meter alone if you already have ISM electrodes, or choose a bundled kit suited to your application.
Pro (S7-Series) meters and kits let you choose from twelve language options and USB connectivity for downloading readings. The brilliant display is sunlight proof—ideal for outdoor use. Additional LED status lights let you make readiness checks at a glance.
1 Year WarrantyIP 67
Specifications
  • DescriptionPortable Conductivity Meter; Pro Field Kit