Mettler Toledo Generator Electrodes for EVA C1 and C3 Karl Fischer Titrators
Plug and play generator electrodes work with EVA coulometric Karl Fischer titrators
  • Constructed of borosilicate-3.3 glass
  • Platinum anode and cathode
  • Measures 2.7 cm dia x 13.8 cm L
773,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
Mettler Toledo 31028002 Generator Electrode with Diaphragm for EVA C1 and C3 Coulometric Karl Fischer Titrators 3333229 Mfr # 31028002
COMPARE
₹773,000.00INR / Each
Mettler Toledo 31028003 Generator Electrode without Diaphragm for EVA V1 and V3 Coulometric Karl Fischer Titrators 3333230 Mfr # 31028003
COMPARE
₹773,000.00INR / Each