Mettler Toledo 31028002 Generator Electrode with Diaphragm for EVA C1 and C3 Coulometric Karl Fischer Titrators
Representative image only.
Mettler Toledo 31028002 Generator Electrode with Diaphragm for EVA C1 and C3 Coulometric Karl Fischer Titrators
Mettler Toledo | Mfr: 31028002| Item: 3333229
Plug and play generator electrodes work with EVA coulometric Karl Fischer titrators
₹773,000.00INR / Each
₹773,000.00INR / Each
Product Description
Key Features
  • Constructed of borosilicate-3.3 glass
  • Platinum anode and cathode
  • Measures 2.7 cm dia x 13.8 cm L
  • Minimizes interference and regulates ion flow
  • Provides enhanced stability and long lifespan
  • Performs automatic exchange of anolyte and manual exchange of catholyte
  • High accuracy for samples with very low amounts of water
  • 1 year warranty
1 Year Warranty
Specifications
  • Special FeaturesWith diaphragm
  • DescriptionGenerator Electrode with Diaphragm for EVA C1 and C3 Coulometric Karl Fischer Titrators