Mettler Toledo 31028003 Generator Electrode without Diaphragm for EVA V1 and V3 Coulometric Karl Fischer Titrators
Representative image only.
Mettler Toledo 31028003 Generator Electrode without Diaphragm for EVA V1 and V3 Coulometric Karl Fischer Titrators
Mettler Toledo | Mfr: 31028003| Item: 3333230
Plug and play generator electrodes work with EVA coulometric Karl Fischer titrators
₹773,000.00INR / Each
₹773,000.00INR / Each
Product Description
Key Features
  • Constructed of borosilicate-3.3 glass
  • Platinum anode and cathode
  • Measures 2.7 cm dia x 13.8 cm L
  • Allows direct solution contact for fast response time and sensitivity
  • Easier to clean than an electrode with diaphragm
  • Performs automatic exchange of anolyte
  • Has a slightly lower accuracy for samples with low amounts of water
  • 1 year warranty
1 Year Warranty
Specifications
  • Special FeaturesWithout diaphragm
  • DescriptionGenerator Electrode without Diaphragm for EVA V1 and V3 Coulometric Karl Fischer Titrators