Mark-10 Motorized Test Stand Wedge Grips
Ideal for tensile testing
  • Jam-free design offers improved gripping
  • Tru slot for samples of unlimited width
  • Ergonomic handle reduces hand fatigue
These heavy duty self-tightening grips come in a capacity range of 50 to 2000 lbF offering you a wide range of tensile testing. The grip design prevents jamming and improves gripping. The smooth and easy operation of the lever reduces hand fatigue.
175,000.00 - ₹214,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mark-10 G1061 Wedge Grip, 0.75” Jaw Width, 5/16-18F 2530447 Mfr # G1061
COMPARE
G1061 Test Stand Wedge Grip, 1200 lb capacity
₹195,000.00INR / Each
Mark-10 G1061-1 Wedge Grip, 0.375” Jaw Width, 5/16-18F 2530448 Mfr # G1061-1
COMPARE
G1061-1 Test Stand Wedge Grip, 200 lb capacity
₹175,000.00INR / Each
Mark-10 G1061-2 Wedge Grip, 1” Jaw Width, 1/2-20F 2530449 Mfr # G1061-2
COMPARE
G1061-2 Test Stand Wedge Grip, 2000 lb capacity
₹214,000.00INR / Each