Mark-10 G1061-2 Wedge Grip, 1” Jaw Width, 1/2-20F

Representative image only.

Mark-10 G1061-2 Wedge Grip, 1” Jaw Width, 1/2-20F

Mark-10 | Mfr: G1061-2| Item: 2530449

Ideal for tensile testing

₹214,000.00INR / Each

₹214,000.00INR / Each
Product Description

Key Features

  • Jam-free design offers improved gripping
  • Tru slot for samples of unlimited width
  • Ergonomic handle reduces hand fatigue
  • Ideal for use with most force gauges and test stands
  • 3 year warranty

More About this Item

These heavy duty self-tightening grips come in a capacity range of 50 to 2000 lbF offering you a wide range of tensile testing. The grip design prevents jamming and improves gripping. The smooth and easy operation of the lever reduces hand fatigue.

3 Year Warranty
Specifications
  • DescriptionG1061-2 Test Stand Wedge Grip, 2000 lb capacity
  • Warranty3 year
Reviews
Q&A