Mark-10 G1061 Wedge Grip, 0.75” Jaw Width, 5/16-18F

Representative image only.

Mark-10 G1061 Wedge Grip, 0.75” Jaw Width, 5/16-18F

Mark-10 | Mfr: G1061| Item: 2530447

Ideal for tensile testing

₹195,000.00INR / Each

₹195,000.00INR / Each
Product Description

Key Features

  • Jam-free design offers improved gripping
  • Tru slot for samples of unlimited width
  • Ergonomic handle reduces hand fatigue
  • Ideal for use with most force gauges and test stands
  • 3 year warranty

More About this Item

These heavy duty self-tightening grips come in a capacity range of 50 to 2000 lbF offering you a wide range of tensile testing. The grip design prevents jamming and improves gripping. The smooth and easy operation of the lever reduces hand fatigue.

3 Year Warranty
Specifications
  • DescriptionG1061 Test Stand Wedge Grip, 1200 lb capacity
  • Warranty3 year
Reviews
Q&A