Labconco Flask Adapters Fast-Freeze® Flasks
Easily connect Fast-Freeze® flasks to freeze dryer valves
Use the adapters to attach Fast-Freeze® flasks to valve ports on Labconco drying chambers and manifolds.
3,600.00 - ₹10,200.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Connections Material Compatible With
Availability Pricing
Labconco FreeZone Flask Adapter, Straight, Glass, 1/2" Dia 0335400 Mfr # 7545000
COMPARE
1/2" diameter Glass Compatible with 40 and 80 mL Fast-Freeze flask tops
₹3,600.00INR / Each
Labconco FreeZone Flask Adapter, Straight, Glass, 3/4" Dia 0335401 Mfr # 7545200
COMPARE
3/4" diameter Glass Compatible with 120, 150, 300, 600, 900, 1200, and 2000 mL Fast-Freeze flask tops
₹4,500.00INR / Each
Labconco FreeZone Flask Adapter, Straight, Glass, 1/2" to 3/4" Dia 0335402 Mfr # 7545400
COMPARE
1/2" to 3/4" diameter Glass Compatible with 40, 80, 120, 150, 300, 600, 900, 1200, and 2000 mL Fast-Freeze flask tops
₹6,400.00INR / Each
Labconco FreeZone Flask Adapter, 45° Elbow, Glass, 1/2" 0335410 Mfr # 7545600
COMPARE
1/2" diameter Glass Compatible with 40 and 80 mL Fast-Freeze flask tops
₹6,300.00INR / Each
Labconco FreeZone Flask Adapter, 45° Elbow, Glass, 3/4" 0335411 Mfr # 7545800
COMPARE
3/4" diameter Glass Compatible with 120, 150, 300, 600, 900, 1200, and 2000 mL Fast-Freeze flask tops
₹6,300.00INR / Each
Labconco FreeZone Flask Adapter, 45° Elbow, Glass, 1/2" to 3/4" 0335412 Mfr # 7546000
COMPARE
1/2" to 3/4" diameter Glass Compatible with 40, 80, 120, 150, 300, 600, 900, 1200, and 2000 mL Fast-Freeze flask tops
₹10,200.00INR / Each
Labconco FreeZone Flask Adapter, Straight, SS, 1/2" Dia 0335430 Mfr # 7547000
COMPARE
1/2" diameter Stainless steel Compatible with 40 and 80 mL Fast-Freeze flask tops
₹8,900.00INR / Each
Labconco FreeZone Flask Adapter, Straight, SS, 3/4" Dia 0335431 Mfr # 7547200
COMPARE
3/4" diameter Stainless steel Compatible with 120, 150, 300, 600, 900, 1200, and 2000 mL Fast-Freeze flask tops
₹8,900.00INR / Each
Showing 1- 8 of 8