Labconco FreeZone Flask Adapter, Straight, SS, 3/4" Dia
Representative image only.
Labconco FreeZone Flask Adapter, Straight, SS, 3/4" Dia
Labconco | Mfr: 7547200| Item: 0335431
Easily connect Fast-Freeze® flasks to freeze dryer valves
₹8,900.00INR / Each
₹8,900.00INR / Each
Product Description
Key Features
  • Stainless steel construction
  • Straight style
  • 3/4" diameter
  • Compatible with 120, 150, 300, 600, 900, 1200, and 2000 mL Fast-Freeze Flask Tops
  • 1 year warranty
More About this Item
Use the adapters to attach Fast-Freeze® flasks to valve ports on Labconco drying chambers and manifolds.
1 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithCompatible with 120, 150, 300, 600, 900, 1200, and 2000 mL Fast-Freeze flask tops
  • Connections3/4" diameter
  • DescriptionFlask Adapter, Straight, SS, 3/4" Dia