Labconco FreeZone Flask Adapter, Straight, Glass, 1/2" Dia
Representative image only.
Labconco FreeZone Flask Adapter, Straight, Glass, 1/2" Dia
Labconco | Mfr: 7545000| Item: 0335400
Easily connect Fast-Freeze® flasks to freeze dryer valves
₹3,600.00INR / Each
₹3,600.00INR / Each
Product Description
Key Features
  • Borosilicate glass construction
  • Straight style
  • 1/2" diameter
  • Compatible with 40 and 80 mL Fast-Freeze Flask Tops
  • 1 year warranty
More About this Item
Use the adapters to attach Fast-Freeze® flasks to valve ports on Labconco drying chambers and manifolds.
1 Year Warranty
Specifications
  • MaterialGlass
  • Compatible WithCompatible with 40 and 80 mL Fast-Freeze flask tops
  • Connections1/2" diameter
  • DescriptionFlask Adapter, Straight, Glass, 1/2" Dia