Labconco® Base Storage Cabinets
Provide support for your fume hood while adding additional storage
  • Choose between four types of storage cabinets – Acid, Solvent, Standard, and Vacuum Pump
  • Constructed of powder-coated steel
  • Supports loads of up to 800 pounds
Standard base cabinets are ideal for general storage of small laboratory equipment, mild acids ,and other reagents.
Acid storage cabinets allow you to safely store and vent acids and other corrosive liquids. The interior is constructed with a corrosion-resistant polyethylene liner to enhance corrosion resistance.
Solvent storage cabinets are suitable for storing and venting solvents and other flammable liquids. These cabinets will protect the interior contents in case of an external fire. These cabinets are constructed with dual wall powder-coated steel and feature a 1.5” air gap for fire protection.
Vacuum pump storage cabinets are utilized for vacuum pump storage.
Work surfaces are sold separately and required. Spillstopper work surfaces keep accidental spills and liquids away from the seam between the hood wall and surrounding countertop.
382,000.00 - ₹957,000.00INR / Each
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Hood Width (in) Description
Availability Pricing
Labconco 8075000 Variable height bench, with locking casters 0909963 Mfr # 8075000
COMPARE
Variable height bench, with locking casters
₹471,000.00INR / Each
Labconco 9900300 Standard Base Storage Cabinet for 24" Fume Hood, Right Hinge 0910900 Mfr # 9900300
COMPARE
35.5 24 Standard Base Storage Cabinet for 24" Fume Hood, Right Hinge
₹382,000.00INR / Each
Labconco 9900600 Standard Base Storage Cabinet for 24" Fume Hood, Left Hinge 0910910 Mfr # 9900600
COMPARE
35.5 24 Standard Base Storage Cabinet for 24" Fume Hood, Left Hinge
₹382,000.00INR / Each
Labconco 9900200 Standard Base Storage Cabinet for 30" Fume Hood 0911500 Mfr # 9900200
COMPARE
36.75 30 Standard Base Storage Cabinet for 30" Fume Hood
₹400,000.00INR / Each
Labconco 9903200 Solvent Base Storage Cabinet for 30" Fume Hood 0911502 Mfr # 9903200
COMPARE
36.75 30 Solvent Base Storage Cabinet for 30" Fume Hood
₹848,000.00INR / Each
Labconco 9901200 Acid Base Storage Cabinet for 30" Fume Hood 0911504 Mfr # 9901200
COMPARE
36.75 30 Acid Base Storage Cabinet for 30" Fume Hood
₹478,000.00INR / Each
Labconco 9900000 Standard Base Storage Cabinet for 48" Fume Hood 0911506 Mfr # 9900000
COMPARE
36.75 48 Standard Base Storage Cabinet for 48" Fume Hood
Call For Price
Labconco 9903000 Solvent Base Storage Cabinet for 48" Fume Hood 0911508 Mfr # 9903000
COMPARE
36.75 48 Solvent Base Storage Cabinet for 48" Fume Hood
Call For Price
Labconco 9901000 Acid Base Storage Cabinet for 48" Fume Hood 0911509 Mfr # 9901000
COMPARE
36.75 48 Acid Base Storage Cabinet for 48" Fume Hood
₹719,000.00INR / Each
Labconco 9901100 Acid Base Storage Cabinet for 36" Fume Hood 0911524 Mfr # 9901100
COMPARE
36.75 36 Acid Base Storage Cabinet for 36" Fume Hood
₹496,000.00INR / Each
Labconco 9902100 Solvent Base Storage Cabinet for 36" Fume Hood 0911526 Mfr # 9902100
COMPARE
36.75 36 Solvent Base Storage Cabinet for 36" Fume Hood
₹665,000.00INR / Each
Labconco 9900100 Standard Base Storage Cabinet for 36" Fume Hood 0911528 Mfr # 9900100
COMPARE
36.75 36 Standard Base Storage Cabinet for 36" Fume Hood
₹412,000.00INR / Each
Labconco 9904700 ADA-Compliant Standard Base Storage Cabinet for 36" Fume Hood 0911551 Mfr # 9904700
COMPARE
32.75 36 ADA-Compliant Standard Base Storage Cabinet for 36" Fume Hood
Call For Price
Labconco 9904800 ADA-Compliant Standard Base Storage Cabinet for 48" Fume Hood 0911552 Mfr # 9904800
COMPARE
32.75 48 ADA-Compliant Standard Base Storage Cabinet for 48" Fume Hood
Call For Price
Labconco 9905400 ADA-Compliant Acid Base Storage Cabinet for 30" Fume Hood 0911553 Mfr # 9905400
COMPARE
32.75 30 ADA-Compliant Acid Base Storage Cabinet for 30" Fume Hood
Call For Price
Labconco 9905500 ADA-Compliant Acid Base Storage Cabinet for 36" Fume Hood 0911554 Mfr # 9905500
COMPARE
32.75 36 ADA-Compliant Acid Base Storage Cabinet for 36" Fume Hood
₹681,000.00INR / Each
Labconco 9905600 ADA-Compliant Acid Base Storage Cabinet for 48" Fume Hood 0911555 Mfr # 9905600
COMPARE
32.75 48 ADA-Compliant Acid Base Storage Cabinet for 48" Fume Hood
₹839,000.00INR / Each
Labconco 9906400 ADA-Compliant Solvent Base Storage Cabinet for 30" Fume Hood 0911556 Mfr # 9906400
COMPARE
32.75 30 ADA-Compliant Solvent Base Storage Cabinet for 30" Fume Hood
₹884,000.00INR / Each
Labconco 9906500 ADA-Compliant Solvent Base Storage Cabinet for 36" Fume Hood 0911557 Mfr # 9906500
COMPARE
32.75 36 ADA-Compliant Solvent Base Storage Cabinet for 36" Fume Hood
Call For Price
Labconco 9906600 ADA-Compliant Solvent Base Storage Cabinet for 48" Fume Hood 0911558 Mfr # 9906600
COMPARE
32.75 48 ADA-Compliant Solvent Base Storage Cabinet for 48" Fume Hood
₹957,000.00INR / Each
Showing 1 - 20 of 22