Labconco 8075000 Variable height bench, with locking casters
Representative image only.
Labconco 8075000 Variable height bench, with locking casters
Labconco | Mfr: 8075000| Item: 0909963
Provide support for your fume hood while adding additional storage
₹471,000.00INR / Each
₹471,000.00INR / Each
Product Description
Key Features
  • Choose between four types of storage cabinets – Acid, Solvent, Standard, and Vacuum Pump
  • Constructed of powder-coated steel
  • Supports loads of up to 800 pounds
  • 1 year warranty
More About this Item
Standard base cabinets are ideal for general storage of small laboratory equipment, mild acids ,and other reagents.
Acid storage cabinets allow you to safely store and vent acids and other corrosive liquids. The interior is constructed with a corrosion-resistant polyethylene liner to enhance corrosion resistance.
Solvent storage cabinets are suitable for storing and venting solvents and other flammable liquids. These cabinets will protect the interior contents in case of an external fire. These cabinets are constructed with dual wall powder-coated steel and feature a 1.5” air gap for fire protection.
Vacuum pump storage cabinets are utilized for vacuum pump storage.
Work surfaces are sold separately and required. Spillstopper work surfaces keep accidental spills and liquids away from the seam between the hood wall and surrounding countertop.
1 Year Warranty
Specifications
  • DescriptionVariable height bench, with locking casters