Labconco 9900600 Standard Base Storage Cabinet for 24" Fume Hood, Left Hinge

Representative image only.

Labconco 9900600 Standard Base Storage Cabinet for 24" Fume Hood, Left Hinge

Labconco | Mfr: 9900600| Item: 0910910

Provide support for your fume hood while adding additional storage

₹382,000.00INR / Each

₹382,000.00INR / Each
Product Description

Key Features

  • Choose between four types of storage cabinets – Acid, Solvent, Standard, and Vacuum Pump
  • Constructed of powder-coated steel
  • Supports loads of up to 800 pounds
  • 1 year warranty

More About this Item

Standard base cabinets are ideal for general storage of small laboratory equipment, mild acids ,and other reagents.

Acid storage cabinets allow you to safely store and vent acids and other corrosive liquids. The interior is constructed with a corrosion-resistant polyethylene liner to enhance corrosion resistance.

Solvent storage cabinets are suitable for storing and venting solvents and other flammable liquids. These cabinets will protect the interior contents in case of an external fire. These cabinets are constructed with dual wall powder-coated steel and feature a 1.5” air gap for fire protection.

Vacuum pump storage cabinets are utilized for vacuum pump storage.

Work surfaces are sold separately and required. Spillstopper work surfaces keep accidental spills and liquids away from the seam between the hood wall and surrounding countertop.

1 Year WarrantyConformite EuropeenneETL ListedISO9001:2000 Certified Supplier
Specifications
  • Hood Width (in)24
  • Exterior Width (in)24
  • Exterior Height (in)35.5
  • Exterior Depth (in)22
  • Interior Width (in)24
  • DescriptionStandard Base Storage Cabinet for 24" Fume Hood, Left Hinge
  • Warranty1 year
Reviews
Q&A