Kruss Glass Polarimeter Tubes
Fill with optically active solution to measure optical rotation
  • Glass construction
  • Without temperature control
Measure the optical rotation of polarized light as it passes through an optically active substance.
113,000.00 - ₹142,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (mm) Special Features Tube Length (mm)
Availability Pricing
Kruss PRG-100 Glass Polarimeter Tube, Standard, 100 mm L 8125041 Mfr # PRG-100
COMPARE
None 100
₹113,000.00INR / Each
Kruss PRG-200 Glass Polarimeter Tube, Standard, 200 mm L 8125042 Mfr # PRG-200
COMPARE
None 200
₹120,000.00INR / Each
Kruss PRG-100-E Glass Polarimeter Tube, Funnel Fill, 100 mm L 8125043 Mfr # PRG-100-E
COMPARE
Filling funnel 100
₹119,000.00INR / Each
Kruss PRG-200-E Glass Polarimeter Tube, Funnel-Fill, 200 mm L 8125044 Mfr # PRG-200-E
COMPARE
Filling funnel 200
₹142,000.00INR / Each