Koflo Tube Mixers
Mix fluids as you pump them through a pipe line
  • Able to mix light viscosity to high viscosity components
  • Accept standard tube fittings
These mixers consist of a series of fixed right- and left-hand elements. Tube and mixing elements are made of 316 stainless steel—ideal for mixing high-viscosity fluids such as epoxies. Mixers have plain ends and accept standard tube fittings,
25,300.00 - ₹44,100.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mixer Length (in) Mounting Type No. Of Elements
Availability Pricing
Koflo 3/16-17 316 SS tube mixer; 3/16" OD; 17 elements 0466950 Mfr # 3/16-17
COMPARE
4.875 0.1875" plain end 17
₹25,300.00INR / Each
Koflo 3/16-21 316 SS tube mixer; 3/16" OD; 21 elements 0466952 Mfr # 3/16-21
COMPARE
6 0.1875" plain end 21
₹25,700.00INR / Each
Koflo 3/16-27 316 SS tube mixer; 3/16" OD; 27 elements 0466954 Mfr # 3/16-27
COMPARE
7.5 0.1875" plain end 27
₹44,100.00INR / Each
Koflo 1/4-21 316 SS Tube Mixer, 21 Elements; 1/4" OD 0466956 Mfr # 1/4-21
COMPARE
7 0.25" plain end 21
₹25,800.00INR / Each
Koflo 1/4-27 316 SS Tube Mixer, 27 Elements; 1/4" OD 0466958 Mfr # 1/4-27
COMPARE
9.25 0.25" plain end 27
₹28,600.00INR / Each
Koflo 1/4-34 316 SS Tube Mixer, 34 Elements; 1/4" OD 0466960 Mfr # 1/4-34
COMPARE
11.5 0.25" plain end 34
₹31,000.00INR / Each
Koflo 3/8-21 316 SS Tube Mixer, 21 Elements; 3/8" OD 0466962 Mfr # 3/8-21
COMPARE
11 0.375" plain end 21
₹26,100.00INR / Each
Koflo 3/8-27 316 SS Tube Mixer, 27 Elements; 3/8" OD 0466964 Mfr # 3/8-27
COMPARE
14 0.375" plain end 27
₹29,100.00INR / Each
Koflo 3/8-32 316 SS Tube Mixer, 32 Elements; 3/8" OD 0466966 Mfr # 3/8-32
COMPARE
17 0.375" plain end 32
₹31,000.00INR / Each
Koflo 1/2-15 316 SS Tube Mixer, 15 Elements; 1/2" OD 0466968 Mfr # 1/2-15
COMPARE
11.875 0.5" plain end 15
₹31,700.00INR / Each
Koflo 1/2-21 316 SS Tube Mixer, 21 Elements; 1/2" OD 0466970 Mfr # 1/2-21
COMPARE
16.375 0.5" plain end 21
₹35,300.00INR / Each
Koflo 1/2-32 316 SS Tube Mixer, 32 Elements; 1/2" OD 0466972 Mfr # 1/2-32
COMPARE
24.75 0.5" plain end 32
₹39,100.00INR / Each
Showing 1- 12 of 12