Koflo 3/16-17 316 SS tube mixer; 3/16" OD; 17 elements
Representative image only.
Koflo 3/16-17 316 SS tube mixer; 3/16" OD; 17 elements
Koflo | Mfr: 3/16-17| Item: 0466950
Mix fluids as you pump them through a pipe line
₹25,300.00INR / Each
₹25,300.00INR / Each
Product Description
Key Features
  • Able to mix light viscosity to high viscosity components
  • Accept standard tube fittings
  • Limited Lifetime Warranty
More About this Item
These mixers consist of a series of fixed right- and left-hand elements. Tube and mixing elements are made of 316 stainless steel—ideal for mixing high-viscosity fluids such as epoxies. Mixers have plain ends and accept standard tube fittings,
Limited Lifetime Warranty
Specifications
  • Material316 Stainless Steel
  • No. Of Elements17
  • Pipe Id (in)0.13
  • Pipe Id (mm)3.302
  • Mixer Length (in)4.875
  • Mounting Type0.1875" plain end
  • Description316 SS tube mixer; 3/16" OD; 17 elements
  • WarrantyLTD Life Time