Koflo 1/4-21 316 SS Tube Mixer, 21 Elements; 1/4" OD
Representative image only.
Koflo 1/4-21 316 SS Tube Mixer, 21 Elements; 1/4" OD
Koflo | Mfr: 1/4-21| Item: 0466956
Mix fluids as you pump them through a pipe line
₹25,800.00INR / Each
₹25,800.00INR / Each
Product Description
Key Features
  • Able to mix light viscosity to high viscosity components
  • Accept standard tube fittings
  • Limited Lifetime Warranty
More About this Item
These mixers consist of a series of fixed right- and left-hand elements. Tube and mixing elements are made of 316 stainless steel—ideal for mixing high-viscosity fluids such as epoxies. Mixers have plain ends and accept standard tube fittings,
Limited Lifetime Warranty
Specifications
  • Material316 Stainless Steel
  • No. Of Elements21
  • Pipe Id (in)0.19
  • Pipe Id (mm)4.826
  • Mixer Length (in)7
  • Mounting Type0.25" plain end
  • Description316 SS Tube Mixer, 21 Elements; 1/4" OD
  • WarrantyLTD Life Time