Koflo PVDF Schedule 80 In-Line Pipe Mixers
Mix fluids as you pump them through a pipe line
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.
48,000.00 - ₹116,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mixer Length (in) Mounting Type No. Of Elements
Availability Pricing
Koflo 1/2-80-10-6-2 Pipe Mixer, Kynar schedule 80, 1/2" NPT(M), 6 elements 0466928 Mfr # 1/2-80-10-6-2
COMPARE
5 0.5" NPT(M) 6
₹48,000.00INR / Each
Koflo 1/2-80-10-12-2 Pipe Mixer, Kynar schedule 80, 1/2" NPT(M), 12 elements 0466929 Mfr # 1/2-80-10-12-2
COMPARE
10 0.5" NPT(M) 12
₹82,400.00INR / Each
Koflo 3/4-80-10-6-2 Pipe Mixer, Kynar schedule 80, 3/4" NPT(M), 6 elements 0466930 Mfr # 3/4-80-10-6-2
COMPARE
6 0.75" NPT(M) 6
₹52,200.00INR / Each
Koflo 1.5-80-10-6-2 Pipe Mixer, Kynar schedule 80, 1-1/2" NPT(M), 6 elements 0466938 Mfr # 1.5-80-10-6-2
COMPARE
12 1.5" NPT(M) 6
₹69,400.00INR / Each
Koflo 2-80-10-6-2 Pipe Mixer, Kynar schedule 80, 2" NPT(M), 6 elements 0466942 Mfr # 2-80-10-6-2
COMPARE
16 2" NPT(M) 6
₹87,300.00INR / Each
Koflo 2-80-10-12-2 Pipe Mixer, Kynar schedule 80, 2" NPT(M), 12 elements 0466944 Mfr # 2-80-10-12-2
COMPARE
31 2" NPT(M) 12
₹116,000.00INR / Each