Koflo 3/4-80-10-6-2 Pipe Mixer, Kynar schedule 80, 3/4" NPT(M), 6 elements
Representative image only.
Koflo 3/4-80-10-6-2 Pipe Mixer, Kynar schedule 80, 3/4" NPT(M), 6 elements
Koflo | Mfr: 3/4-80-10-6-2| Item: 0466930
Mix fluids as you pump them through a pipe line
₹52,200.00INR / Each
₹52,200.00INR / Each
Product Description
Key Features
  • Limited Lifetime Warranty
More About this Item
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.
Limited Lifetime Warranty
Specifications
  • MaterialPVDF
  • No. Of Elements6
  • Pipe Id (in)0.82
  • Pipe Id (mm)20.83
  • Mixer Length (in)6
  • Mounting Type0.75" NPT(M)
  • DescriptionPipe Mixer, Kynar schedule 80, 3/4" NPT(M), 6 elements
  • WarrantyLTD Life Time