Koflo 1/2-80-10-12-2 Pipe Mixer, Kynar schedule 80, 1/2" NPT(M), 12 elements
Representative image only.
Koflo 1/2-80-10-12-2 Pipe Mixer, Kynar schedule 80, 1/2" NPT(M), 12 elements
Koflo | Mfr: 1/2-80-10-12-2| Item: 0466929
Mix fluids as you pump them through a pipe line
₹82,400.00INR / Each
₹82,400.00INR / Each
Product Description
Key Features
  • Limited Lifetime Warranty
More About this Item
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.
Limited Lifetime Warranty
Specifications
  • MaterialPVDF
  • No. Of Elements12
  • Pipe Id (in)0.546
  • Pipe Id (mm)13.87
  • Mixer Length (in)10
  • Mounting Type0.5" NPT(M)
  • DescriptionPipe Mixer, Kynar schedule 80, 1/2" NPT(M), 12 elements
  • WarrantyLTD Life Time