Kimcote Safety-Coated Heavy-Walled Glass Filtering Flasks
KimCote™ plastic coating for added safety
Heavy-wall borosilicate glass offers mechanical strength for vacuum filtration. Flask has a #2 side arm for connection to 1⁄4" (6 mm) ID flexible tubing. Autoclavable to 120°C (250°F).
22,500.00 - ₹68,600.00INR / Case of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Stopper Number Accepted
Availability Pricing
DWK Life Sciences (Kimble) 953760-0122 Heavy-Wall Safety-Coated Glass Filtering Flask, 125 mL; 1/Cs WW-34524-49 Mfr # 953760-0122
COMPARE
125 Glass 5
₹22,500.00INR / Case of 1
DWK Life Sciences (Kimble) 953760-1002 Heavy-Wall Safety-Coated Glass Filtering Flask, 1000 mL; 1/Cs WW-34524-51 Mfr # 953760-1002
COMPARE
1000 Glass 8
₹38,700.00INR / Case of 1
DWK Life Sciences (Kimble) 953760-2002 Heavy-Wall Safety-Coated Glass Filtering Flask, 2000 mL; 1/Cs WW-34524-53 Mfr # 953760-2002
COMPARE
2000 Glass 8
₹56,300.00INR / Case of 1
DWK Life Sciences (Kimble) 953760-4002 Heavy-Wall Safety-Coated Glass Filtering Flask, 4000 mL; 1/Cs WW-34524-55 Mfr # 953760-4002
COMPARE
4000 Glass 8
₹68,600.00INR / Case of 1