Kimble Disposable Glass Serological Pipettes
Pipettes have negative graduations to increase versatility
These pipettes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class B glass requirements. They are calibrated ''to deliver'' (TD) with blow-out after free flow of liquid has ended.
40,100.00 - ₹90,900.00INR / Case of 400
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Max Volume (µL) Sterile
Availability Pricing
DWK Life Sciences (Kimble) 72120-11100 Disposable Glass Serological Pipettes, 1 mL, TD; 1000/Cs (Bulk) WW-25553-10 Mfr # 72120-11100
COMPARE
±0.030mL 1000 No
₹66,700.00INR / Case of 1000
DWK Life Sciences (Kimble) 72120-21100 Disposable Glass Serological Pipettes, 2 mL, TD; 500/Cs (Bulk) WW-25553-11 Mfr # 72120-21100
COMPARE
±0.060mL 2000 No
₹50,000.00INR / Case of 500
DWK Life Sciences (Kimble) 72120-5110 Disposable Glass Serological Pipettes, 5 mL, TD; 500/Cs (Bulk) WW-25553-12 Mfr # 72120-5110
COMPARE
±0.150mL 5000 No
₹53,500.00INR / Case of 500
DWK Life Sciences (Kimble) 72120-10110 Disposable Glass Serological Pipettes, 10 mL, TD; 500/Cs (Bulk) WW-25553-13 Mfr # 72120-10110
COMPARE
±0.030mL 10000 No
₹59,600.00INR / Case of 500
DWK Life Sciences (Kimble) 72100-11100 Sterile Plugged Glass Serological Pipettes, 1mL, TD; 1000/Cs (Bulk) WW-25553-14 Mfr # 72100-11100
COMPARE
±0.030mL 1000 Yes
₹77,300.00INR / Case of 1000
DWK Life Sciences (Kimble) 72100-21100 Sterile Plugged Glass Serological Pipettes, 2mL, TD; 500/Cs (Bulk) WW-25553-15 Mfr # 72100-21100
COMPARE
±0.060mL 2000 Yes
₹40,100.00INR / Case of 500
DWK Life Sciences (Kimble) 72100-5110 Sterile Plugged Glass Serological Pipettes, 5mL, TD; 500/Cs (Bulk) WW-25553-16 Mfr # 72100-5110
COMPARE
±0.150mL 5000 Yes
₹54,100.00INR / Case of 500
DWK Life Sciences (Kimble) 72100-10110 Sterile Plugged Glass Serological Pipettes, 10mL, TD; 500/Cs (Bulk) WW-25553-17 Mfr # 72100-10110
COMPARE
±0.100mL 10000 Yes
₹76,000.00INR / Case of 500
DWK Life Sciences (Kimble) 72115-11100 Sterile Shorty  Plugged Glass Pipettes, 1mL, TD; 400/Cs (Indv) WW-25553-18 Mfr # 72115-11100
COMPARE
±0.040mL 1000 Yes
₹65,800.00INR / Case of 400
DWK Life Sciences (Kimble) 72115-5110 Sterile Shorty  Plugged Glass Pipettes, 5mL, TD; 200/Cs (Indv) WW-25553-19 Mfr # 72115-5110
COMPARE
±0.200mL 5000 Yes
₹90,900.00INR / Case of 200
DWK Life Sciences (Kimble) 72115-10210 Sterile Shorty  Plugged Glass Pipettes, 10mL, TD; 200/Cs (Indv) WW-25553-20 Mfr # 72115-10210
COMPARE
±0.400mL 10000 Yes
₹89,700.00INR / Case of 200
Showing 1- 11 of 11