Keck Ramp Tubing Clamps
  • Roll wheel forward to restrict tubing and decrease flow.
  • Roll wheel back to open tubing and increase flow.
Unique wheel-in-groove clamp design permits one-handed flow control. Simply remove wheel to insert tubing, then roll wheel back into clamp to your desired flow rate. Made of pure PBT, these clamps are safe to 392°F
(200°C). All four sizes accept tubing with a maximum wall thickness of 3/32" (2.4 mm).
9,100.00 - ₹22,200.00INR / Pkg of 12
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Clamp Type Max Grip Range (InchShort) Description
Availability Pricing
Keck Ramp Clamp Tubing Clamps, 6 mm, 12/Pk 0683503 Mfr # MAKE-UP
COMPARE
Tubing 0.25 Ramp Clamp Tubing Clamps, 6 mm, 12/Pk
₹9,100.00INR / Pkg of 12
Keck Ramp Clamp Tubing Clamps, 10 mm, 12/Pk 0683507 Mfr # MAKE-UP
COMPARE
Tubing 0.375 Ramp Clamp Tubing Clamps, 10 mm, 12/Pk
₹14,700.00INR / Pkg of 12
Keck Ramp Clamp Tubing Clamps, 14 mm, 12/Pk 0683510 Mfr # MAKE-UP
COMPARE
Tubing 0.563 Ramp Clamp Tubing Clamps, 14 mm, 12/Pk
₹11,800.00INR / Pkg of 12
Keck Ramp Tubing Clamp Assortment Kit , 32 Piece 0684180 Mfr # MAKE-UP
COMPARE
Tubing Ramp Tubing Clamp Assortment Kit , 32 Piece
₹22,200.00INR / Each