Keck Ramp Clamp Tubing Clamps, 6 mm, 12/Pk
Representative image only.
Keck Ramp Clamp Tubing Clamps, 6 mm, 12/Pk
Keck | Mfr: MAKE-UP| Item: 0683503
₹9,100.00INR / Pkg of 12
₹9,100.00INR / Pkg of 12
Product Description
Key Features
  • Roll wheel forward to restrict tubing and decrease flow.
  • Roll wheel back to open tubing and increase flow.
More About this Item
Unique wheel-in-groove clamp design permits one-handed flow control. Simply remove wheel to insert tubing, then roll wheel back into clamp to your desired flow rate. Made of pure PBT, these clamps are safe to 392°F
(200°C). All four sizes accept tubing with a maximum wall thickness of 3/32" (2.4 mm).
Specifications
  • Clamp MaterialPolybutylene Terephthalate (PBT)
  • Clamp TypeTubing
  • ColorYellow
  • Max Grip Range (InchShort)0.25
  • DescriptionRamp Clamp Tubing Clamps, 6 mm, 12/Pk