Keck Ramp Tubing Clamp Assortment Kit , 32 Piece
Representative image only.
Keck Ramp Tubing Clamp Assortment Kit , 32 Piece
Keck | Mfr: MAKE-UP| Item: 0684180
₹22,200.00INR / Each
₹22,200.00INR / Each
Product Description
Key Features
  • Roll wheel forward to restrict tubing and decrease flow.
  • Roll wheel back to open tubing and increase flow.
More About this Item
Unique wheel-in-groove clamp design permits one-handed flow control. Simply remove wheel to insert tubing, then roll wheel back into clamp to your desired flow rate. Made of pure PBT, these clamps are safe to 392°F
(200°C). All four sizes accept tubing with a maximum wall thickness of 3/32" (2.4 mm).
Specifications
  • Clamp MaterialPolybutylene Terephthalate (PBT)
  • Clamp TypeTubing
  • DescriptionRamp Tubing Clamp Assortment Kit , 32 Piece