Katanax® Crucibles for Katanax® Fluxers
Quality crucibles provide excellent temperature uniformity for reliable results
  • Feature straight walls
  • 30 mL capacity
  • Measures 32 mm H x 40.5 mm OD
Select crucibles made from 95% Pt-5% Au (borate fusion) or 100 % zirconium (peroxide fusion). Platinum/gold crucibles are available with or without flared top. Flared top adds rigidity and enhances the pouring action.
Select heavy models with thicker walls and bottom for greater durability, longer lifespan, and better temperature control.
708,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Description
Availability Pricing
Katanax Crucible, 95% Pt-5% Au, Straight, Regular, 26 g 0457837 Mfr # KP1001A
COMPARE
95% Pt-5% Au Crucible, 95% Pt-5% Au, Straight, Regular, 26 g
Call For Price
Katanax Crucible, 95% Pt-5% Au, Straight, Heavy, 30 g 0457838 Mfr # KP1002A
COMPARE
95% Pt-5% Au Crucible, 95% Pt-5% Au, Straight, Heavy, 30 g
Call For Price
Katanax Crucible, 95% Pt-5% Au, Flared, Regular, 28 g 0457853 Mfr # KP1101A
COMPARE
95% Pt-5% Au Crucible, 95% Pt-5% Au, Flared, Regular, 28 g
Call For Price
Katanax Crucible, 95% Pt-5% Au, Flared, Heavy, 43 g 0457858 Mfr # KP1112A
COMPARE
95% Pt-5% Au Crucible, 95% Pt-5% Au, Flared, Heavy, 43 g
Call For Price
Katanax Crucible, 95% Pt-5% Au, Flared, Regular, 26 g 0477625 Mfr # KP1102A
COMPARE
95% Pt-5% Au Crucible, 95% Pt-5% Au, Flared, Regular, 26 g
₹708,000.00INR / Each