Capacity (mL)
CA Origin
Katanax
Katanax
Katanax® has been creating high-quality electric fusion equipment for over a decade. Our fully automated electric fluxers are designed to prepare glass disks (beads) for XRF analysis, or solutions for AA and ICP, including peroxide and pyrosulfate fusions. Our fluxers combine exceptional fusion accuracy with all the advantages of electric power, speed, simplicity, and a robust design for the experienced or novice user.