IKA Big Squid Stirrers with Unique Motifs
Big brother to the IKA Mini Squid
  • Offers same features with larger stirring capacity up to 1.5L
  • Your choice of motifs
This variable speed stirrer is ideal for low-viscosity fluids. Glass top plate and Hytrel® base offer excellent chemical resistance.
13,400.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
High Speed (rpm) Max Temperature (° F) Top Plate Length (in)
Availability Pricing
IKA 3050002 Big Squid Magnetic Stirrer, Froggy 1594925 Mfr # 3050002
COMPARE
Call For Price
IKA 3050001 Big Squid Magnetic Stirrer, Star 1594926 Mfr # 3050001
COMPARE
₹13,400.00INR / Each
IKA 3050005 Big Squid Magnetic Stirrer, maleon 1594927 Mfr # 3050005
COMPARE
Call For Price
IKA 3050004 Big Squid Magentic Stirrer, Ocean 1594928 Mfr # 3050004
COMPARE
Call For Price
IKA 3573000 Big Squid mag Pineapple 1594929 Mfr # 3573000
COMPARE
Call For Price
IKA 3573200 Big Squid MAG Strawberry 1594930 Mfr # 3573200
COMPARE
Call For Price
IKA 3573400 Big Squid Magnetic Stirrer, Kiwi 1594931 Mfr # 3573400
COMPARE
Call For Price
IKA 3573600 Big Squid Magnetic Stirrer, Orange 1594932 Mfr # 3573600
COMPARE
Call For Price
Showing 1- 8 of 8