IKA 3573000 Big Squid mag Pineapple
Representative image only.
IKA 3573000 Big Squid mag Pineapple
IKA | Mfr: 3573000| Item: 1594929
Big brother to the IKA Mini Squid
Call For Price
Call For Price
Product Description
Includes
2-m cord.
Key Features
  • Offers same features with larger stirring capacity up to 1.5L
  • Your choice of motifs
More About this Item
This variable speed stirrer is ideal for low-viscosity fluids. Glass top plate and Hytrel® base offer excellent chemical resistance.
Specifications
  • No. Of Stirring Positions1
  • DescriptionBig Squid mag Pineapple