IKA 3050002 Big Squid Magnetic Stirrer, Froggy
Representative image only.
IKA 3050002 Big Squid Magnetic Stirrer, Froggy
IKA | Mfr: 3050002| Item: 1594925
Big brother to the IKA Mini Squid
Call For Price
Call For Price
Product Description
Includes
2-m cord.
Key Features
  • Offers same features with larger stirring capacity up to 1.5L
  • Your choice of motifs
More About this Item
This variable speed stirrer is ideal for low-viscosity fluids. Glass top plate and Hytrel® base offer excellent chemical resistance.
Specifications
  • No. Of Stirring Positions1
  • DescriptionBig Squid Magnetic Stirrer, Froggy