IDEX® Quick-Stop Inline Luer Chromatography Check Valves
Check valve protection with luer connection convenience
  • Add back-flow protection to your system
  • Simply disconnect luer fitting to stop the flow
Quick-Stop Luer Check Valves provide the fast connect/disconnect without the mess and hazard of spills. Connect the valve to your inline tubing. Check valve opens automatically once the luer connection is engaged. Disconnect the luer union to close the valve and stop the flow.
29,800.00 - ₹36,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description Inlet Port Outlet Port
Availability Pricing
Idex Quick-Stop Inline Check Valve, Luer (F) to 1/4-28 (F) Flat Bottom; 1/EA 0201426 Mfr # P-697
COMPARE
Quick-Stop Inline Check Valve, Luer (F) to 1/4-28 (F) Flat Bottom; 1/EA 1/4-28 (F) flat bottom Luer (F)
₹29,800.00INR / Each
Idex Quick-Stop Inline Check Valve Assembly, 1/4-28 (F) Flat Bottom to Luer to 1/4-28 (F) Flat Bottom; 1/EA 0201599 Mfr # P-696
COMPARE
Quick-Stop Inline Check Valve Assembly, 1/4-28 (F) Flat Bottom to Luer to 1/4-28 (F) Flat Bottom; 1/EA 1/4-28 (F) flat botom [to Luer (F)] [Luer (M) to] 1/4-28 (F) flat bottom
₹36,300.00INR / Each
Idex Quick-Stop Inline Check Valve, Bulkhead, Luer (F) to 1/4-28 (M) Flat Bottom; 1/EA 0201601 Mfr # P-699
COMPARE
Quick-Stop Inline Check Valve, Bulkhead, Luer (F) to 1/4-28 (M) Flat Bottom; 1/EA 1/4-28 (F) flat bottom Luer (F)
₹32,900.00INR / Each