Idex Quick-Stop Inline Check Valve, Luer (F) to 1/4-28 (F) Flat Bottom; 1/EA
Representative image only.
Idex Quick-Stop Inline Check Valve, Luer (F) to 1/4-28 (F) Flat Bottom; 1/EA
Idex | Mfr: P-697| Item: 0201426
Check valve protection with luer connection convenience
₹29,800.00INR / Each
₹29,800.00INR / Each
Buy It Together
Current Item
Idex Quick-Stop Inline Check Valve, Luer (F) to 1/4-28 (F) Flat Bottom; 1/EA
Idex Quick-Stop Inline Check Valve, Luer (F) to 1/4-28 (F) Flat Bottom; 1/EA
0201426
₹29,800.00INR / Each
+ =
Total Price 34900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Add back-flow protection to your system
  • Simply disconnect luer fitting to stop the flow
  • Luer (F) to 1/4-28 (F) flat-bottom connections
  • 1 month warranty
More About this Item
Quick-Stop Luer Check Valves provide the fast connect/disconnect without the mess and hazard of spills. Connect the valve to your inline tubing. Check valve opens automatically once the luer connection is engaged. Disconnect the luer union to close the valve and stop the flow.
Check valve is made of PEEK with gold-plated stainless steel spring. Order luer adapter 02014-12 for a complete luer connection.
1 Month Warranty
Specifications
  • Outlet PortLuer (F)
  • Connection TypeUNF x Luer
  • Inlet Port1/4-28 (F) flat bottom
  • Wetted MaterialsPEEK/SS
  • DescriptionQuick-Stop Inline Check Valve, Luer (F) to 1/4-28 (F) Flat Bottom; 1/EA