Idex Quick-Stop Inline Check Valve Assembly, 1/4-28 (F) Flat Bottom to Luer to 1/4-28 (F) Flat Bottom; 1/EA
Representative image only.
Idex Quick-Stop Inline Check Valve Assembly, 1/4-28 (F) Flat Bottom to Luer to 1/4-28 (F) Flat Bottom; 1/EA
Idex | Mfr: P-696| Item: 0201599
Check valve protection with luer connection convenience
₹36,300.00INR / Each
₹36,300.00INR / Each
Buy It Together
Current Item
Idex Quick-Stop Inline Check Valve Assembly, 1/4-28 (F) Flat Bottom to Luer to 1/4-28 (F) Flat Bottom; 1/EA
Idex Quick-Stop Inline Check Valve Assembly, 1/4-28 (F) Flat Bottom to Luer to 1/4-28 (F) Flat Bottom; 1/EA
0201599
₹36,300.00INR / Each
+ =
Total Price 41400.00 ADD 2 ITEMS TO CART
Product Description
Includes
One P-697 Quick Stop check valve, one P-655 adapter
Key Features
  • Add back-flow protection to your system
  • Simply disconnect luer fitting to stop the flow
  • Complete assembly with luer connections for fast connect/disconnect
  • 1 year warranty
More About this Item
Quick-Stop Luer Check Valves provide the fast connect/disconnect without the mess and hazard of spills. Connect the valve to your inline tubing. Check valve opens automatically once the luer connection is engaged. Disconnect the luer union to close the valve and stop the flow.
Check valve assembly is comprised of check valve 02014-26 and adapter 02014-12. Check valve is made of PEEK with gold-plated stainless steel spring. Adapter is made of PEEK.
1 Year Warranty
Specifications
  • Outlet Port[Luer (M) to] 1/4-28 (F) flat bottom
  • Connection TypeUNF x Luer
  • Inlet Port1/4-28 (F) flat botom [to Luer (F)]
  • Wetted MaterialsPEEK/SS
  • DescriptionQuick-Stop Inline Check Valve Assembly, 1/4-28 (F) Flat Bottom to Luer to 1/4-28 (F) Flat Bottom; 1/EA