Idex® Micro-Metering Valves

Excellent for fine control of fluid flow rates
  • Easily control fluid output for a wide range of low flow applications
Micro-metering valves can reduce outgoing flow to as low as 3.5 µL/min. Use valves in peristaltic pump fluid transfer applications, mass spectroscopy, fraction collection, and helium sparging lines. Valves have 0.020" (0.5 mm) thru holes.
15,100.00 - ₹17,900.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Thread size UNF(M) (port 2) (in) Description
Availability Pricing
Idex Micro-Metering Valve With Fittings, 10-32 Coned, 1/16" OD Tubing WW-02013-90 Mfr # P-446
COMPARE
1/16 10-32 coned Micro-Metering Valve With Fittings, 10-32 Coned, 1/16" OD Tubing; 1/EA
₹17,900.00INR / Each
Idex Micro-Metering Valve With Fittings, 1/4-28 Flat-Bottom, 1/16" OD Tubing; 1/EA WW-02016-90 Mfr # P-445
COMPARE
1/16 1/4-28 flat-bottom Micro-Metering Valve With Fittings, 1/4-28 Flat-Bottom, 1/16" OD Tubing; 1/EA
₹17,800.00INR / Each
Idex Micro-Metering Valve without Fittings, 10-32 Coned; 1/Ea WW-02016-92 Mfr # P-446NF
COMPARE
10-32 coned Micro-Metering Valve Without Fittings, 10-32 Coned; 1/EA
₹15,100.00INR / Each
Idex Micro-Metering Valve With Fittings, 1/4-28 Flat-Bottom, 1/8" OD Tubing; 1/EA WW-02016-93 Mfr # P-447
COMPARE
1/8 1/4-28 flat-bottom Micro-Metering Valve With Fittings, 1/4-28 Flat-Bottom, 1/8" OD Tubing; 1/EA
₹17,100.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat