Idex Micro-Metering Valve With Fittings, 10-32 Coned, 1/16" OD Tubing
Representative image only.
Idex Micro-Metering Valve With Fittings, 10-32 Coned, 1/16" OD Tubing
Idex | Mfr: P-446| Item: 0201390
Excellent for fine control of fluid flow rates
₹24,000.00INR / Each
₹24,000.00INR / Each
Buy It Together
Current Item
Idex Micro-Metering Valve With Fittings, 10-32 Coned, 1/16" OD Tubing
Idex Micro-Metering Valve With Fittings, 10-32 Coned, 1/16" OD Tubing
0201390
₹24,000.00INR / Each
+ =
Total Price 36900.00 ADD 2 ITEMS TO CART
Product Description
Includes
PEEK fittings.
Key Features
  • Easily control fluid output for a wide range of low flow applications
  • Includes 10-32 coned fittings for 1/16" OD tubing
  • 1 month warranty
More About this Item
Micro-metering valves can reduce outgoing flow to as low as 3.5 µL/min. Use valves in peristaltic pump fluid transfer applications, mass spectroscopy, fraction collection, and helium sparging lines. Valves have 0.020" (0.5 mm) thru holes.
Valve has 10-32 coned ports, comes with one-piece PEEK fittings, and has 0.020" (0.5 mm) thru holes.
Volume listed is maximum volume with valve fully open.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK
  • Valve MaterialPEEK, PTFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)10-32 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Volume (µL)7.2
  • ColorBlack
  • DescriptionMicro-Metering Valve With Fittings, 10-32 Coned, 1/16" OD Tubing; 1/EA